missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDA2R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550400
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total EDA2R protein.
EDA2R Polyclonal antibody specifically detects EDA2R in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
EDA2R | |
Polyclonal | |
Unconjugated | |
EDA2R | |
9430060M22Rik; ectodysplasin A2 isoform receptor; ectodysplasin A2 receptor; EDA2R; EDA-A2 receptor; EDAA2R; EDA-A2R; RGD1564025; Tnfrsf27; Tumor necrosis factor receptor superfamily member 27; tumor necrosis factor receptor superfamily member XEDAR; UNQ2448/PRO5727/PRO34080; X linked ectodysplasin receptor; Xedar; X-linked ectodysplasin-A2 receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60401 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9HAV5 | |
EDA2R | |
Fusion protein corresponding to residues near the C terminal of human ectodysplasin A2 receptor | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |