Läs mer
Tocris Bioscience™ Edaglitazone
Potent and selective PPARγ agonist; antidiabetic
Brand: Tocris Bioscience™ 4784/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Edaglitazone is a potent and selective PPARγ agonist (EC50 values are 35.6 and 1053 nM for PPARγ and PPARα cofactor recruitment respectively. Enhances insulin sensitivity in obese, but not lean, rats. Antidiabetic and orally bioavailable.Specifikationer
| Edaglitazone | |
| 5-[[4-[2-(5-Methyl-2-phenyl-4-oxazolyl)ethoxy]benzo[b]thien-7-yl]methyl]-2,4-thiazolidinedione | |
| Soluble to 100mM in DMSO | |
| Bioactive Small Molecule |
| 213411-83-7 | |
| 10 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use