Learn More
EDC4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559461
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VERALETRHE QEQRRLERAL AEGQQRGGQL QEQLTQQLSQ ALSSAVAGRL ERSIRDEIKK TVPPCVSRSL EPMAGQLSNS VATKLTAVEG SMKENISKLL KSKNLTDA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
EDC4 Polyclonal antibody specifically detects EDC4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
EDC4 | |
Polyclonal | |
Unconjugated | |
EDC4 | |
Autoantigen Ge-1; autoantigen RCD8; autoantigen RCD-8; BC022641; Edc4; enhancer of mRNA decapping 4; enhancer of mRNA-decapping protein 4; fe26b05; GE1; Ge-1; HEDL5; HEDLS; Human enhancer of decapping large subunit; LRRG00115; Nrn1l; RCD8; RCD-8; RGD1309136; si:ch211-237c7.1; si:dkey-16n13.6; wu:fe26b05 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23644 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P2E9 | |
EDC4 | |
Recombinant protein corresponding to Human EDC4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only