missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDEM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26226-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
EDEM1 belongs to the glycosyl hydrolase 47 family. It extracts misfolded glycoproteins, but not glycoproteins undergoing productive folding, from the calnexin cycle. It is directly involved in endoplasmic reticulum-associated degradation (ERAD) and targets misfolded glycoproteins for degradation in an N-glycan-dependent manner.Specifications
EDEM1 | |
Polyclonal | |
Unconjugated | |
EDEM1 | |
EDEM, EDEM1, KIAA0212 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192193, 297504, 9695 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q925U4, Q92611 | |
EDEM1 | |
EDEM1 Fusion Protein Ag24046 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |