missing translation for 'onlineSavingsMsg'
Learn More

EDEM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26226-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16814745

  • 4721.67 SEK / 150µL
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

EDEM1 belongs to the glycosyl hydrolase 47 family. It extracts misfolded glycoproteins, but not glycoproteins undergoing productive folding, from the calnexin cycle. It is directly involved in endoplasmic reticulum-associated degradation (ERAD) and targets misfolded glycoproteins for degradation in an N-glycan-dependent manner.
Specifications

Specifications

EDEM1
Polyclonal
Unconjugated
EDEM1
EDEM, EDEM1, KIAA0212
Rabbit
Antigen Affinity Chromatography
RUO
192193, 297504, 9695
-20°C
Liquid
Immunoprecipitation, Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q925U4, Q92611
EDEM1
EDEM1 Fusion Protein Ag24046
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers