missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Edetate Disodium U.S.P., Macron Fine Chemicals™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly
Specifications
Packaging | POLYSTORMOR |
Quantity | 500 g |
IUPAC Name | disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
Formula Weight | 372.24 |
Grade | USP |
Chemical Name or Material | Edetate Disodium |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'