missing translation for 'onlineSavingsMsg'
Learn More

Edetate Disodium U.S.P., Macron Fine Chemicals™

macron

Brand:  Macron Fine Chemicals 1395-06

Additional Details : CAS Number : 6381-92-6 Weight : 2.50000kg

Packaging Poly
Quantity 2.5kg

 View more versions of this product

Product Code. 15567904

Quantity Price
1 3823.00 SEK / 2.50kg
6 3589.03 SEK / 2.50kg
Estimated Shipment
21-01-2021
Add to basket


Description

Description

Specifications

Chemical Identifiers

6381-92-6
372.238
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
C10H18N2Na2O10
OVBJJZOQPCKUOR-UHFFFAOYSA-L
44120005
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]

Specifications

Edetate Disodium
2.5kg
C10H18N2Na2O10
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
44120005
USP
Poly
6381-92-6
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]
372.238
372.24
Videos
SDS
Documents