missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ EDG6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523221
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence homology: 100% homologous in human; mouse/rat (95%).
EDG6 (S1P4) belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids. The ligand for EDG6 (S1P4) is sphingosine-1-phosphate. There are 8 known members of the EDG receptor family and they are implicated in mediating growth related effects such as induction of cellular proliferation, alterations in differentiation and survival and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis. EDG receptors are developmentally regulated and differ in tissue distribution. They couple to multiple types of G proteins to signal through ras and MAP kinase, rho, phospholipase C and several protein tyrosine kinases. EDG6 (S1P4) is expressed in lymphoid and hematopoietic tissue and in the lungs. Recently, the designation of the EDG receptors has been modified to include the ligand for the receptor. Thus, the new designation for the EDG6 (S1P4) receptor is the sphingosine 1-phosphate receptor-4 (S1P4).
Specifikationer
| EDG6 | |
| Polyclonal | |
| Unconjugated | |
| S1PR4 | |
| Edg6; EDG-6; Endothelial differentiation G-protein coupled receptor 6; endothelial differentiation G-protein-coupled receptor 6; endothelial differentiation, G protein coupled receptor 6; endothelial differentiation, G-protein-coupled receptor 6; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 6; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 6; LPC1; Lysophospholipid receptor C1; S1P receptor 4; S1P receptor Edg-6; S1P(4) receptor; S1p4; S1PR4; SLP4; sphingosine 1-phosphate receptor 4; Sphingosine 1-phosphate receptor Edg-6; sphingosine-1-phosphate receptor 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13611, 8698 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.0 mg/mL | |
| PBS with 0.05% sodium azide | |
| O95977, Q9Z0L1 | |
| S1PR4 | |
| A portion of amino acids 1-50 of human Sphingolipid Receptor Edg6/S1P4. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering