Description
- Personal noise dosimeter offers advanced technology packaged in a cable-free, compact frame for a lighter, more ergonomic way to monitor noise levels
- Rechargeable lithium polymer battery
- Thin, contoured mount hugs the shoulder; compact frame offers superior comfort
Specifications
Specifications
| Batteritid | 60 timmar |
| Certifikat och efterlevnad | CE, IEC 61252, ANSI S1.25 |
| Inkluderar | Docking Station |
| Mätområde | Dose, Exposure, Identity Field, Lavg/Leq, Max, Min, Peak, Run Time, SEL (Lep), SPL, Threshold, TWA, Upper Limit (UL) |