missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell EDGE™ 48-126 Polyurethane Coated Seamless Knit Industrial Work Gloves
Click to view available options
Size:
6
7
8
9
10
11
Description
- An inexpensive, light duty glove with quality features
- Seamless structure ensures comfort and encourages safety
- 15-gauge polyester offers flexibility and dexterity
- High tactility in dry or slightly oily environments
- Dark color hides dirt for an extended useful life
Specifications
ANSI Abrasion Rating | ANSI 4 |
Construction | Seamless Knit |
Coating Material | Polyurethane |
Color | Black |
Hand | Specific |
Material | Polyester |
Product Type | Seamless Knit Gloves |
Size | 8 |
For Use With (Application) | Automotive Assembly, Oil/Gas, and Machinery/Equipment Operation |
Glove Cuff Style | Knit |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'