Läs mer
Ansell EDGE™ 48-706 Medium Duty Cut-Resistant Gloves
Produktkod. 18822985 Handla allt Ansell Produkter
Change view
Storlek:
7
8
9
10
11
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Storlek
18822985 7
18832985 8
18842985 9
18852985 10
18862985 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 18822985 Leverantör Ansell Leverantörsnummer 48706070
  • ONLINE EXCLUSIVE PRICE
    16291.30 SEK
    11348.83 SEK / Paket med 144
    Save 4942.47 SEK
    30% Av
Beräknad leverans: 02-09-2026
Lägg i varukorgen
Lägg i varukorgen
Seamless knit gloves with palm coated nitrile are ideal for medium-duty applications requiring a high level of cut performance in dry or slightly oily environments.
  • High durability
  • Nitrile foam coating delivers abrasion resistance and grip
  • Breathable liner allows high level of comfort for end users
  • Secure grip in dry and slightly oily environments
  • Made of a seamless liner,
  • Alternative to heavy duty leather and polyurethane palm dipped cut resistant knitted gloves
  • Uses:
    • Automotive, construction, glass manufacturing, machinery, equipment, metal fabrication
  • TRUSTED_SUSTAINABILITY
    Specifikationer
    ANSI Abrasion Rating ANSI 5
    Certifieringar/efterlevnad (Europa) CE, EN 420:2003, A1:2009, Cat II
    Konstruktion Knit
    Beläggningsmaterial Nitrile Foam
    Färg Gray, Black
    Betyg för skärmotstånd ANSI A3
    Produkttyp Cut-resistant Gloves
    Storlek 7
    EN388 Standard EN 388:4X43C
    För användning med (applikation) General maintenance, hardware assembly, machining, stamping, cutting, molding, plastic injections, raw material handling, seat and frame build
    Handske manschettstil Knit
    Liner material HPPE, Fiberglass, Nylon, Polyester, Spandex
    Modell 48706070
    Kvantitet 12 Pairs/Pk., 12 Pk./Cs.
    Serie EDGE™ 48-706
    Längd (engelska) 8.6 in.
    Längd (metrisk) 22 cm