Läs mer
Edoxaban 4-Carboxylic Acid Hydrochloride, TRC
Produktkod. 30376236
Change view
Kvantitet:
1 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30376236 25 mg
30375710 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30376236 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE55557025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 834919-19-6
Molekylformel C22H25ClN6O5S . xHCl
Molekylvikt (g/mol) 521
Synonym (1S,3R,4S)-4-[[2-[(5-Chloro-2-pyridinyl)amino]-2-oxoacetyl]amino]-3-[[(4,5,6,7-tetrahydro-5-methylthiazolo[5,4-c]pyridin-2-yl)carbonyl]amino]cyclohexanecarboxylic Acid, (1S,3R,4S)-4-[[[(5-chloro-2-pyridinyl)amino]oxoacetyl]amino]-3-[[(4,5,6,7-tetrahydro-5-methylthiazolo[5,4-c]pyridin-2-yl)carbonyl]amino]cyclohexanecarboxylic Acid
LEDER CN1CCc2nc(sc2C1)C(=O)N[C@@H]3C[C@H](CC[C@@H]3NC(=O)C(=O)Nc4ccc(Cl)cn4)C(=O)O
Specifikationer
Kemiskt namn eller material Edoxaban 4-Carboxylic Acid Hydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C22H25ClN6O5S/c1-29-7-6-14-16(10-29)35-21(27-14)20(32)26-15-8-11(22(33)34)2-4-13(15)25-18(30)19(31)28-17-5-3-12(23)9-24-17/h3,5,9,11,13,15H,2,4,6-8,10H2,1H3,(H,25,30)(H,26,32)(H,33,34)(H,24,28,31)/t11-,13-,15+/m0/s1
Formel vikt 520.1296