missing translation for 'onlineSavingsMsg'
Learn More

EDTA Disodium Salt Solution, 0.01M, Chem-Lab

Used in electrophoresis and molecular biology applications. EDTA Disodium Salt Solution, 0.01M, Chem-Lab is a chelator of divalent cations for use in TH titrations. Solution contains 3.723g EDTA diNa • 2H2O per liter of water (0.01 M).

Brand:  Chem Lab CL05.0501.5000

Additional Details : Weight : 5.00000kg

Quantity 5L

 View more versions of this product

Product Code. 11993143

  • 550.00 SEK / 5 L

Estimated Shipment: 11-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

EDTA Disodium Salt Dihydrate is a chelator of divalent cations. It inhibits enzymes, such as metalloproteases, that require divalent cations for activity. It is also used in many enzyme buffers and as an enzyme inactivator at high concentrations.
Specifications

Chemical Identifiers

6381-92-6
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
44120005
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
C10H18N2Na2O10
MFCD00150037,MFCD00003541
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate

Specifications

0.01mol/L (± 0.00004, 20°C)
Odorless
Colorless
5L
6381-92-6
MFCD00150037,MFCD00003541
Soluble
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
372.24
372.24g/mol
1g/cm3
100°C
0°C
4 to 5
C10H18N2Na2O10
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
44120005
Liquid
Videos
SDS
Documents
Certificates