Läs mer
Hanna Instruments™ Educational pH/ORP Meter
Produktkod. 10585611
Mått (L x B x H):
185 x 82 x 53 mm
Förpackningsstorlek:
Varje
Produktkod. 10585611
Brand: Hanna Instruments™ HI8014
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Ideal for education and field applications. Hanna Instruments™ Educational pH/ORP Meter is a rugged, handheld pH/ORP meter specifically designed with ease of use in mind. This affordable, portable meter is perfect for teaching students the fundamentals of pH and ORP measurement.
HI8014 Educational pH/ORP Meter performs pH measurements with manual temperature compensation. It also performs ORP measurements using the mV range and optional ORP electrode (HI3131B). Two-point calibration can be performed with trimmers on the front panel.
  • Manual Temperature Compensation
  • Two-point Calibration
  • Low Battery Indicator
  • Optional ORP Electrode
Specifikationer
Inkluderar HI1230B pH electrode, 1m cable, calibration screwdriver, battery and instructions
Noggrannhet (mV) ±1
Noggrannhet (pH) ±0.01
För användning med (applikation) Ideal for education and field applications
Kalibrering Manual via Trimmers
Område (pH) 0 to 14
Upplösning (pH) 0.01
Räckvidd (mV) ±1999
Upplösning (mV) 1mV
Kalibreringspunkter 2
Temperaturområde (metriskt) 0°C to 100°C
Temperaturområde (engelska) 32°F to 212°F
Mått (L x B x H) 185 x 82 x 53 mm
Vikt (metrisk) 265 g
Vikt (engelska) 9.3 oz.
Garanti 2 Years (6 Months Probe)
Sondtyp HI1230B PEI body pH electrode with BNC connector
Detektionsområde 0 to 14 pH
Temperaturkompensation 0°C to 100°C, Manual
Temperatur (engelska) Drift 32°F to 122°F
Temperatur (metrisk) Drift 0°C to 50°C
Batterityp 9V
Typ Educational Portable pH
Batteritid 100 hr.