Learn More
Edwards Vacuum Rotary Vane Vacuum Pump Gas Ballast Oil Return Kit
Conserve pump oil, by returning oil trapped in mist filter to the pump and allows extended operation of pump with gas ballast
Brand: Edwards A50523000
Additional Details : Weight : 3.00000kg
Description
The RV Gas Ballast Oil Drain Kits continuously return the oil trapped in the EMF Oil Mist Filter to the pump. When fitted, the pump will operate as if the gas ballast control is in position II (high flow position). Kit helps in reduced oil loss from the pump and minimizes the need to check the pump oil-level and to refill the pump with oil.
- Adjusable gas ballast flow rate
Order Info
EMF oil mist filter and pump sold separately
Specifications
Model RV3, RV5, RV8, RV12 pump | |
Gas Ballast Oil Kit |