missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16091-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody can recognize both EEF1A1 and EEF1A2.
This gene encodes an isoform of the alpha subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This isoform (alpha 2) is expressed in brain, heart and skeletal muscle, and the other isoform (alpha 1) is expressed in brain, placenta, lung, liver, kidney, and pancreas. This gene may be critical in the development of ovarian cancer. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
EEF1A2 | |
Polyclonal | |
Unconjugated | |
EEF1A2 | |
eEF1A 2, EEF1A2, EEF1AL, EF 1 alpha 2, EF1A, Elongation factor 1 alpha 2, HS1, Statin S1, STN, STNL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13628, 1917, 24799 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62631, P62632, Q05639 | |
EEF1A2 | |
EEF1A2 Fusion Protein Ag6875 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |