missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1B2 Mouse anti-Human, Clone: 3E1C11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60329-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a translation elongation factor. The protein is a guanine nucleotide exchange factor involved in the transfer of aminoacylated tRNAs to the ribosome. Alternative splicing results in three transcript variants which differ only in the 5′ UTR.Specifications
EEF1B2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P24534 | |
EEF1B2 | |
EEF1B2 Fusion Protein Ag0135 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot, Immunofluorescence | |
3E1C11 | |
Unconjugated | |
EEF1B2 | |
EEF1B, EEF1B1, EEF1B2, EF 1 beta, EF1B, Elongation factor 1 beta | |
Mouse | |
Protein G | |
RUO | |
1933 | |
-20°C | |
Liquid |