Learn More
EEF1B2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557123
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FGDLKSPAGL QVLNDYLADK SYIEGYVPSQ ADVAVFEAVS SPPPADLCHA LRWYNHIKSY EKEKASLPGV KKALGKYGPA DVEDTTGSGA TD Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
EF-1 (elongation factor-1) is a multi-protein complex that is responsible forthe delivery of aminoacyl-tRNAs to the ribosome. The EF-1 protein complexis composed of four subunits: EF-1 α, EF-1 β, EF-1 δ and EF-1 γ. EF-1 β is a225 amino acid member of the EF-1 β/EF-1 δ protein family and contains oneGST C-terminal domain. Both EF-1 β and EF-1 δ stimulate the exchange ofGDP bound to EF-1 α to GTP. Phosphorylation of EF-1 β affects the GDP/GTP exchange rate. Alternative splicing events of the gene that encodes EF-1 β result in three transcript variants that differ only in the 5' UTR.Specifications
EEF1B2 | |
Polyclonal | |
Unconjugated | |
EEF1B2 | |
2810017J07Rik; EEF1B; EEF1B1; EEF1B2; EF1B; EF-1-beta; Elongation factor 1-beta; eukaryotic translation elongation factor 1 beta 1; eukaryotic translation elongation factor 1 beta 2; eukaryotic translation elongation factor 1-beta homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1933 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P24534 | |
EEF1B2 | |
Recombinant protein corresponding to Human EEF1B2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only