Learn More
EFCAB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554813
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: YCFEVFDLNG DGFISKEEMF HMLKNSLLKQ PSEEDPDEGI KDLVEITLKK MDHDHDGKLS FADYELAVRE ETLLLEAFGP CLPDPKSQME FEAQVFKDPN Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
EFCAB1 Polyclonal antibody specifically detects EFCAB1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
EFCAB1 | |
Polyclonal | |
Unconjugated | |
EFCAB1 | |
EFCAB1; EF-hand calcium binding domain 1; EF-hand calcium-binding domain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79645 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HAE3 | |
EFCAB1 | |
Recombinant protein corresponding to Human EFCAB1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only