Learn More
EFCAB12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558064
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NPASQPQAPP KPIPSFKVLE ARDIQEQPED RKTWLSQRSK LRQELESFGD VKRWLENKPS ITPSEAKVLH MIHEEQSAQP NASQATTRTT Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 52%.
EFCAB12 Polyclonal antibody specifically detects EFCAB12 in Human samples. It is validated for Immunohistochemistry.Specifications
EFCAB12 | |
Polyclonal | |
Unconjugated | |
EFCAB12 | |
C3orf25; EFCAB12; EF-hand calcium binding domain 12; EF-hand calcium-binding domain-containing protein 12; EF-hand domain-containing protein C3orf25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90288 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NXP0 | |
EFCAB12 | |
Recombinant protein corresponding to Human EFCAB12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only