missing translation for 'onlineSavingsMsg'
Läs mer

PeproTech EGF Polyclonal Antibody, PeproTech®, Invitrogen™

Produktkod. 17853993
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
100 μg
50 μg
Produktkod. Kvantitet
17853993 100 μg
17825063 50 μg
17864973 1 mg
3 options
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 17853993

Brand: PeproTech 500P174G100UG

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Goat Polyclonal Antibody

AA Sequence of recombinant protein: NSYPGCPSSY DGYCLNGGVC MHIESLDSYT CNCVIGYSGD RCQTRDLRWW ELR. Preparation: Produced from sera of goats immunized with highly pure Recombinant Murine EGF. Anti-Murine EGF-specific antibody was purified by affinity chromatography employing an immobilized Murine EGF matrix. Sandwich ELISA: To detect Murine EGF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine EGF (500-P174GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine EGF. Western Blot: To detect Murine EGF by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine EGF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen EGF
Användningsområden ELISA, Western Blot
Klassificering Polyclonal
Koncentration 0.1-1.0 mg/mL
Konjugera Unconjugated
Formulering PBS with no preservative
Gen EGF
Genaccessionsnr. P01132
Gene Alias AI790464; beta-urogastrone; EGF; Epidermal growth factor; epidermal growth factor (beta-urogastrone); epidermal growth factor precursor; H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); sb:eu639; URG; Urogastrone
Gensymboler EGF
Värd art Goat
Immunogen E.coli-derived Recombinant Murine EGF.
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 13645
Målarter Mouse
Innehåll och lagring -20°C
Produkttyp Antibody
Form Lyophilized
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.