Learn More
EGFL5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565602
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: SELEPECDQCK DGYIGPNCNK CENGYYNFDS ICRKCQCHGH VDPVKTPKIC KPESGECINC LHNTTGFWCE NCLEGYVHDL EGNCIKKEVI LPTPEGSTIL VSNASLTTSV PTPVINSTFT PTTLQTIFSV ST Highest antigen sequence identity to the following orthologs - mouse 86%, rat 84%.
EGFL5 Polyclonal antibody specifically detects EGFL5 in Human samples. It is validated for Immunocytochemistry.Specifications
EGFL5 | |
Polyclonal | |
Unconjugated | |
MEGF9 | |
4933405H16Rik; 9630025C07; EGFL5; EGF-like protein 5; EGF-like-domain, multiple 5; Epidermal growth factor-like protein 5; KIAA0818; Megf9; mKIAA0818; multiple EGF like domains 9; multiple EGF-like domains protein 9; multiple EGF-like-domains 9; multiple epidermal growth factor-like domains protein 9; UNQ671/PRO1305 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1955 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H1U4 | |
MEGF9 | |
Recombinant Human EGFL5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only