Learn More
Abnova™ EGFL7 Recombinant Protein
Human EGFL7 full-length ORF recombinant protein with GST-tag at N-terminal
Brand: Abnova™ H00051162-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Theoretical MW (kDa): 53.24
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Best use within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH12377.1 | |
Solution | |
51162 | |
EGFL7 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EGFL7 | |
Human | |
Yes |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
53.24 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
YRPGRRVCAVRAHGDPVSESFVQRVYQPFLTTCDGHRACSTYRTIYRTAYRRSPGLAPARPRYACCPGWKRTSGLPGACGAAICQPPCRNGGSCVQPGRCRCPAGWRGDTCQSDVDERSARRGGCPQRCINTAGSYWCQCWEGHSLSADGTLCVPKGGPPRVAPNPTGVDSAMKEEVQRLQSRVDLLEEKLQLVLAPLHSLASQALEHGLPDPGSLLVHSFQQLGRIDSLSEQISFLEEQLGSCSCKKDS | |
MGC111117/RP11-251M1.2/VE-STATIN/ZNEU1 | |
EGFL7 | |
Wheat Germ (in vitro) | |
GST |