Learn More
EGR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55117-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16826575
  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021
Description
Description
The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.
Specifications
Specifications
EGR1
0.29 mg/mL
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3
P08046, P18146
AT225, early growth response 1, EGR 1, EGR1, G0S30, KROX 24, KROX24, NGFI A, TIS8, Transcription factor ETR103, Transcription factor Zif268, ZIF 268, Zinc finger protein 225, Zinc finger protein Krox 24, ZNF225
Rabbit
IgG
150 μL
-20°C
Primary
13653, 1958
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
EGR1
Liquid
EGR1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse