missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EHD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11382-2-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the gamma subunit of the catalytic core. Alternatively spliced transcript variants encoding different isoforms have been identified. This gene also has a pseudogene on chromosome 14.Specifications
EHD4 | |
Polyclonal | |
Unconjugated | |
EHD4 | |
EH domain containing 4, EH domain containing protein 4, EHD4, HCA10, HCA11, PAST homolog 4, PAST4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192204, 30844, 98878 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9EQP2, Q9H223 | |
Ehd4 | |
EHD4 Fusion Protein Ag1942 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |