missing translation for 'onlineSavingsMsg'
Learn More

EHD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11382-2-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16818423

  • 4721.67 SEK / 150µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the gamma subunit of the catalytic core. Alternatively spliced transcript variants encoding different isoforms have been identified. This gene also has a pseudogene on chromosome 14.
Specifications

Specifications

EHD4
Polyclonal
Unconjugated
EHD4
EH domain containing 4, EH domain containing protein 4, EHD4, HCA10, HCA11, PAST homolog 4, PAST4
Rabbit
Antigen Affinity Chromatography
RUO
192204, 30844, 98878
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9EQP2, Q9H223
Ehd4
EHD4 Fusion Protein Ag1942
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers