Läs mer
EHMT2-IN-1, MedChemExpress
Produktkod. 16487237
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16487237 5 mg
16463116 25 mg
16454528 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16487237 Leverantör Medchem Express Leverantörsnummer HY111778
MedChemExpress EHMT2-IN-1 is a potent EHMT inhibitor, with IC50s of all <100 nM for EHMT1 peptide, EHMT2 peptide and cellular EHMT2. Used in the research of blood disorder or cancer.
EHMT2-IN-1 is a potent EHMT inhibitor, with IC50s of all <100 nM for EHMT1 peptide, EHMT2 peptide and cellular EHMT2. Used in the research of blood disorder or cancer.
Specifikationer
Kemiskt namn eller material EHMT2-IN-1
CAS 2230849-55-3
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C18H23N7O
Kvantitet 5 mg
Löslighetsinformation DMSO : 55 mg/mL (155.62 mM; Need ultrasonic)
LEDER COC1=C(N2N=CC(CNC)=C2)C=C(NC3=NC(NC)=CC(C)=N3)C=C1
Molekylvikt (g/mol) 353.42
Formel vikt 353.42
Procent renhet 99.9%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death