Läs mer
STEMCELL Technologies EHNA (Hydrochloride)
Beskrivning
- EHNA is a reversible adenosine deaminase inhibitor (IC50 = 1.2 μM in human red blood cells) that also selectively inhibits the cGMP-specific phosphodiesterase (PDE2) (IC50 = 0.8 and 2 μM from human and porcine myocardium, respectively, 3.5 μM in rat hepatocyte, and 5.5 μM in human platelet) (Michie et al.; Podzuweit et al.)
- Maintains pluripotency of human embryonic stem cells in the absence of exogenous cytokines, and blocks directed neuronal differentiation (Burton et al.)
Specifikationer
Specifikationer
| CAS | 58337-38-5 |
| Synonym | NSC 263164; erythro-9-(2-Hydroxy-3-nonyl)adenine |
| Kvantitet | 5 mg |
| Molekylformel | C14H23N5O · HCl |
| Renhet | ≥98% |