missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15276-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Necessary for scanning and involved in initiation site selection. Promotes the assembly of 48S ribosomal complexes at the authentic initiation codon of a conventional capped mRNA.Specifications
EIF1 | |
Polyclonal | |
Unconjugated | |
EIF1 | |
A121, EIF 1, EIF1, EIF1A, ISO1, SUI1, Sui1iso1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10209, 20918, 287703 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P41567, P48024 | |
Eif1 | |
EIF1 Fusion Protein Ag7394 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |