missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF2C3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19692-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to EIF2C3.
Small interfering RNAs (siRNAs) have gained much attention for their powerful ability to suppress gene expression. Introduction of double-stranded RNA (dsRNA), that are homologues in sequence to a gene, has proven to suppress that gene′s expression through a process known as RNA interference (RNAi); this process post-transcriptionally silences a gene through mRNA inhibition or degradation. Both biochemical and genetic studies have led to the identification of two, conserved gene families that are universal components of the interference process. These are the Dicer family, which is comprised of members of the Ribonuclease III family of enzymes, and the Argonaute gene family. Argonaute proteins addition to their role in RNAi process, may also influence development, such as, stem cell fate determination.Specifications
EIF2C3 | |
Polyclonal | |
Unconjugated | |
AGO3 | |
AGO3, Argonaute 3, Argonaute3, eIF 2C 3, eIF2C 3, EIF2C3, hAgo3, Protein argonaute 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192669, 214150, 313593 | |
-20°C | |
Liquid |
Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CJF9, Q9H9G7 | |
Ago3 | |
Peptide | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |