Läs mer
eIF4A3-IN-2, MedChemExpress
Produktkod. 16489976
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16489976 50 mg 50mg
16484676 100 mg -
16473568 10 mg -
16416297 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16489976 Leverantör Medchem Express Leverantörsnummer HY101785
MedChemExpress eIF4A3-IN-2 is a highly selective and noncompetitive eukaryotic initiation factor 4A-3 (eIF4A3) inhibitor with an IC50 of 110 nM.
eIF4A3-IN-2 is a highly selective and noncompetitive eukaryotic initiation factor 4A-3 (eIF4A3) inhibitor with an IC50 of 110 nM.
Specifikationer
Kemiskt namn eller material eIF4A3-IN-2
CAS 2095677-20-4
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C25H19Br2ClN4O2
Kvantitet 50 mg
Löslighetsinformation DMSO : 100 mg/mL (165.92 mM; Need ultrasonic)
LEDER O=C(C1=CC=C(Br)C=C1)N2CCN(C(C3=C4N(C=C(Br)C=C4)N=C3)=O)C[C@@H]2C5=CC=C(Cl)C=C5
Molekylvikt (g/mol) 602.7
Formel vikt 602.7
Procent renhet 99.51%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death