Learn More
eIF4G2/DAP5 Mouse anti-Human, Unconjugated, Clone: 3C11A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67428-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The eIF4G2/DAP5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human eIF4G2/DAP5. This antibody recognizes human, mouse, rat antigen. The eIF4G2/DAP5 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
eIF4G2/DAP5 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
3C11A3 | |
Unconjugated | |
150 μL | |
1982 | |
IgG2a |