missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4G3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11281-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is thought to be part of the eIF4F protein complex, which is involved in mRNA cap recognition and transport of mRNAs to the ribosome. Interestingly, a microRNA (miR-520c-3p) has been found that negatively regulates synthesis of the encoded protein, and this leads to a global decrease in protein translation and cell proliferation. Therefore, this protein is a key component of the anti-tumor activity of miR-520c-3p.Specifications
EIF4G3 | |
Polyclonal | |
Unconjugated | |
EIF4G3 | |
eIF 4 gamma 3, eIF 4 gamma II, eIF 4G 3, eIF4G 3, EIF4G3, eIF4GII | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230861, 298573, 8672 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43432, Q80XI3 | |
Eif4g3 | |
EIF4G3 Fusion Protein Ag1811 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |