Learn More
EIF5A Mouse anti-Human, Unconjugated, Clone: 2D1B6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67214-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The EIF5A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EIF5A. This antibody recognizes human, mouse, rat antigen. The EIF5A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
EIF5A | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
2D1B6 | |
Unconjugated | |
150 μL | |
1984 | |
IgG1 |