Learn More
EIF5A Mouse anti-Human, Unconjugated, Clone: 2D1B6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67214-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839735
  • 4721.67 SEK / 150µL
Estimated Shipment: 29-04-2024
to see stock.
Description
Description
The EIF5A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EIF5A. This antibody recognizes human, mouse, rat antigen. The EIF5A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
EIF5A
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
2D1B6
Unconjugated
150 μL
1984
IgG1