Läs mer
Tocris Bioscience™ Elacridar hydrochloride
Beskrivning
P-glycoprotein (P-gp/ABCG1) inhibitor. Blocks P-gp-mediated multidrug resistance (MDR) of the cytotoxic drugs doxorubicin (Cat. No. 2252) and vincristine (Cat. No. 1257) in CHRC5 cells. Also inhibits breast cancer resistance protein (BCRP/ABCG2). Orally active.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Elacridar hydrochloride |
| CAS | 143851-98-3 |
| Kvantitet | 10 mg |
| Molekylformel | C34H34ClN3O5 |
| Formel vikt | 600.1 |
| Renhet | >98% |
| Molekylvikt (g/mol) | 600.112 |