Läs mer
Elaidamide, TRC
Produktkod. 30372949
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30372949 50 mg
1 options
Produktkod. 30372949 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE50000050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Elaidamide
CAS 4303-70-2
Molekylformel C18 H35 N O
Kvantitet 50 mg
Synonym 9-Octadecenamide, (9E)- (9CI, ACI), (9E)-9-Octadecenamide (ACI), 9-Octadecenamide, (E)- (ZCI), Elaidamide (6CI, 7CI, 8CI), (E)-9,10-Octadecenamide, Elaidic acid amide, Elaidic amide, Elaidoylamide, trans-9,10-Octadecenamide, trans-9-Octadecenamide
InChI formel InChI=1S/C18H35NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H2,19,20)/b10-9+
LEDER CCCCCCCC\C=C\CCCCCCCC(=O)N
IUPAC-namn (E)-octadec-9-enamide
Molekylvikt (g/mol) 281.48
Formel vikt 281.2719