Läs mer
Elaidic Acid Methyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Elaidic acid methyl ester |
| CAS | 1937-62-8 |
| Molekylformel | C19 H36 O2 |
| Kvantitet | 1 g |
| Synonym | 9-Octadecenoic acid, methyl ester, (E)-, Elaidic acid, methyl ester (6CI,8CI), 9(E)-Octadecenoic acid methyl ester, Methyl 9(E)-octadecenoate, Methyl elaidate, Methyl trans-9-octadecenoate, trans-Oleic acid methyl ester, Elaidic Acid Methyl Ester, 9-Octadecenoic acid, methyl ester, (9E)- |
| InChI formel | InChI=1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10+ |
| LEDER | CCCCCCCC\C=C\CCCCCCCC(=O)OC |
| IUPAC-namn | methyl (E)-octadec-9-enoate |
| Molekylvikt (g/mol) | 296.49 |
| Formel vikt | 296.2715 |