Läs mer
STEMCELL Technologies Elastase
Produktkod. 17159271
Encompass_Preferred
Change view
Kvantitet:
25 mg
100 mg
Förpackningsstorlek:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17159271 25 mg -
17169271 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17159271 Leverantör STEMCELL Technologies Leverantörsnummer 07453
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
For digestion of native elastin
  • Elastase is a serine protease consisting of a single polypeptide chain of 240 amino acid residues and contains four disulfide bridges (Shotton & Hartley)
  • Elastase is synthesized as a zymogen (proelastase) and is converted to the active form by limited trypsin proteolysis at its N-terminal
  • Elastase preferentially cleaves the peptide bond of C-terminal neutral, non-aromatic amino acid residues (Schellenberger et al.) Elastase is frequently used in combination with other proteases such as collagenase and trypsin to digest fibrous tissues because of its ability to digest native elastin
Specifikationer
För användning med (applikation) Digestion of Native Elastin