Läs mer
STEMCELL Technologies Elastase
Beskrivning
- Elastase is a serine protease consisting of a single polypeptide chain of 240 amino acid residues and contains four disulfide bridges (Shotton & Hartley)
- Elastase is synthesized as a zymogen (proelastase) and is converted to the active form by limited trypsin proteolysis at its N-terminal
- Elastase preferentially cleaves the peptide bond of C-terminal neutral, non-aromatic amino acid residues (Schellenberger et al.) Elastase is frequently used in combination with other proteases such as collagenase and trypsin to digest fibrous tissues because of its ability to digest native elastin
Specifikationer
Specifikationer
| För användning med (applikation) | Digestion of Native Elastin |
| Kvantitet | 100 mg |