Läs mer
Kolmi Hopen™ Elasticated SMS Coveralls with Hood
Handla allt Medicom Produkter
Storlek:
Small
Medium
Large
X-Large
2X-Large
Beskrivning
- Hooded model with flap on zipper
- SMS material makes it possible to better limit release of particles (lint-free)
- Internal seams
- Elasticated with thumb loops at wrists and ankles
- Without pockets to avoid risk of falling objects
- Air and water vapor permeable
- Good tear and abrasion resistance
- Individual packaging
Specifikationer
Specifikationer
| Produkttyp | Cleanroom Coveralls |
| Storlek | Small |
| Material | SMS |
| Färg | Azure Blue |
| Disponibel | Single-use |
| Huva typ | Elastic |
| Sterilitet | Non-sterile |
| Certifikat och efterlevnad | ISO 9001, (EU) 2016/425 |
| För användning med (applikation) | Controlled Environments |
| Vikt (metrisk) | 50 g/m2 |
| Visa mer |