Learn More
Fisherbrand™ Electrode for Comet Assay Tank
Helps to identify cell damage. Fisherbrand™ Electrode for Comet Assay Tank uses a positive or negative charge to push and pull molecules through electrophoresis gel. Electrodes are available for separate purchase.
Brand: Fisherbrand™
Additional Details : Weight : 0.20000kg
Description
For use with Comet Assay Tanks- Positive Electrode pulls molecules through gel
- Negative Electrode pushes molecules through gel
Specifications
Electrophoresis | |
Comet Assay Tank | |
Positive Electrode | |
1 Each |