Learn More
Fisherbrand™ Electrode for Comet Assay Tank
Helps to identify cell damage. Fisherbrand™ Electrode for Comet Assay Tank uses a positive or negative charge to push and pull molecules through electrophoresis gel. Electrodes are available for separate purchase.
1287.56 SEK
Specifications
For Use With (Application) | Electrophoresis |
---|---|
For Use With (Equipment) | Comet Assay Tank |
Quantity | 1 Each |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15559901
|
Fisherbrand™
|
Negative Electrode |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15569901
|
Fisherbrand™
|
Positive Electrode |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
For use with Comet Assay Tanks- Positive Electrode pulls molecules through gel
- Negative Electrode pushes molecules through gel
Specifications
Electrophoresis | |
Comet Assay Tank | |
1 Each |