Learn More
Fisherbrand™ Electrode for Comet Assay Tank
Helps to identify cell damage. Fisherbrand™ Electrode for Comet Assay Tank uses a positive or negative charge to push and pull molecules through electrophoresis gel. Electrodes are available for separate purchase.
1287.56 SEK
Specifications
For Use With (Application) Electrophoresis
For Use With (Equipment) Comet Assay Tank
Quantity 1 Each
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
15559901
View Documents Promotion Details
Fisherbrand™
Negative Electrode This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15569901
View Documents Promotion Details
Fisherbrand™
Positive Electrode This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
For use with Comet Assay Tanks
  • Positive Electrode pulls molecules through gel
  • Negative Electrode pushes molecules through gel
Specifications
Specifications
Electrophoresis
Comet Assay Tank
1 Each