Learn More
Hanna Instruments™ Electrode Holder
Product Code. 10390741
Change view
Click to view available options
Type:
Electrode Holder
Unit Size:
Each
Product Code. Type Unit Size
10390741 Electrode Holder Each
1 options
Product Code. 10390741 Supplier Hanna Instruments™ Supplier No. HI76405
  • ONLINE EXCLUSIVE PRICE
    4914.71 SEK
    2260.79 SEK / Paket med 1
    Save 2653.92 SEK
    54% Av
Beräknad leverans: 30-11-2026
Holds up to three electrodes in any position. Hanna Instruments™ Electrode Holder includes adjustable arm position and stable steel base.
Cantilever stand with a large steel, enamel coated base, holds up to three electrodes or temperature probes in any position.
Specifications
Type Electrode Holder
For Use With (Application) Holding electrodes
Material Epoxy Coated Steel Base