Learn More
Ebro Electronic™ TFN 520 / 530 Robust Thermocouple Thermometer Series
High accurate thermometers for a whole range of measurement and control applications in the laboratory and in industry.
Brand: Ebro Electronic™ 1340-5522A
Additional Details : Weight : 0.16650kg
Description
Temp. Range: -200 to 1200°C (K), -100 to 800°C (J), -50 to 300°C (T), -100 to 600°C (E)Specifications
EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012 | |
Single-use | |
38 mm | |
190 mm | |
TFN 520-SMP | |
0.1°C at −200° to +250°C (K); 0.1°C at −100° to +190°C (J); 0.1°C at −50° to +220°C (T); 0.1°C at −100° to +150°C (E); 1°C for the rest | |
1-channel Thermometer | |
2192°F | |
-328°F | |
-200°C to +1200°C (K), -100°C to +800°C (J), -50°C to +300°C (T), -100°C to +600°C (E) | |
TPN probe series with SMP connector, AG 130, AG 140, AN 141, AN 150 | |
Lithium 3 V/Typ CR 2477 | |
LCD | |
IP40 |
115 x 54 x 22 mm | |
Applications: Heating technology, Air-conditioning, ventilation, Air, gases, Fluids, Motors, engines, Plastic materials, Laboratory | |
Thermometer, calibration certificate, instruction manual | |
Plastic | |
SMP Connector | |
0.3 kg | |
None | |
1200°C | |
-200°C | |
Factory Calibrated | |
Up to 5 years | |
±0.3°C | |
120 mm |