Läs mer
Honeywell™ ELECTROSOFT Class 1 Latex Gloves
Honeywell™ ELECTROSOFT Class 1 Latex Gloves are ideal for electrical work and should be used with leather protectors for mechanical protection.
Brand: Honeywell™ 2091912-10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
- Beige natural latex insulating glove
- Cut edge
- Category RC
- Ergonomic shape and lightly powdered interior make the glove easy to slip on and off
- The natural latex base offers high dielectric characteristics
- Its two-toned composition, with its beige interior/black exterior, indicates wear on the whole of the glove
- Its thickness provides good dexterity while offering protection up to 7500 volts
- To be used with leather protectors for mechanical protection
Specifikationer
EN 60903-2003, CEI 903-2002 | |
Beige | |
1.5 mm |
Latex | |
10 | |
36 cm |
Korrigering av produktinnehåll
Produkttitel
Honeywell™ ELECTROSOFT Class 1 Latex Gloves > Size: 10
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering