Läs mer
Eletriptan hydrobromide, MedChemExpress
Produktkod. 16490698
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16490698 5 mg 5mg
16478258 10 mg -
16454147 50 mg -
16427986 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16490698 Leverantör Medchem Express Leverantörsnummer HYA00105mg
MedChemExpress Eletriptan hydrobromide (Eletriptan HBr) is a selective 5-HT1B and 5-HT1D receptor agonist with Ki of 0.92 nM and 3.14 nM, respectively.
Eletriptan hydrobromide (Eletriptan HBr) is a selective 5-HT1B and 5-HT1D receptor agonist with Ki of 0.92 nM and 3.14 nM, respectively.
Specifikationer
Kemiskt namn eller material Eletriptan hydrobromide
CAS 177834-92-3
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C22H27BrN2O2S
Kvantitet 5 mg
Synonym Eletriptan HBrUK-116044 hydrobromide
Löslighetsinformation DMSO : 110 mg/mL (237.36 mM; Need ultrasonic)
LEDER O=S(CCC1=CC2=C(NC=C2C[C@@H]3N(CCC3)C)C=C1)(C4=CC=CC=C4)=O.Br
Molekylvikt (g/mol) 463.43
Formel vikt 463.43
Procent renhet 98.13%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation