missing translation for 'onlineSavingsMsg'
Learn More

ELMOD1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12761-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840654

  • 4721.67 SEK / 150µL
Estimated Shipment: 29-02-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Acts as a GTPase-activating protein (GAP) toward guanine nucleotide exchange factors like ARL2, ARL3, ARF1 and ARF6, but not for GTPases outside the Arf family.
Specifications

Specifications

ELMOD1
Polyclonal
Unconjugated
ELMOD1
ELMOD1
Rabbit
Antigen Affinity Chromatography
RUO
55531
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N336
ELMOD1
ELMOD1 Fusion Protein Ag3368
150 μL
Primary
Human
Antibody
IgG
Recommendations

Recommendations

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers