missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ELMOD2 Recombinant Protein

Brand:  Abnova™ H00255520-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179683

  • 4515.00 SEK / 10µg
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH15168
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
57.97
8
Glutathione Sepharose 4 Fast Flow
10 ug
MFISLWEFFYGHFFRFWMKWLLRQMTGKCELQRIFDTYVGAQRTHRIENSLTYSKNKVLQKATHVVQSEVDKYVDDIMKEKNINPEKDASFKICMKMCLLQITGYKQLYLDVESVRKRPYDSDNLQHEELLMKLWNLLMPTKKLNARISKQWAEIGFQGDDPKTDFRGMGILGLINLVYFSENYTSEAHQILSRSNHPKLGYSYAIVGTNLTEMAYSLLKSEALKFHLYNLVPGIPTMEHFHQFYCYLVYEFDKFWFEEEPESIMYFNLYREKFHEKIKGLLLDRNVALTLKV
9830169G11Rik/MGC10084
ELMOD2
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
255520
ELMOD2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ELMOD2
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers