Läs mer
Eltrombopag-13C4, TRC
Produktkod. 30137425
Change view
Kvantitet:
1 mg
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137425 25 mg -
30011825 1 mg -
30007993 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137425 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE50800225MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1217230-31-3
Molekylformel C2113C4H22N4O4
Molekylvikt (g/mol) 446.44
Synonym 3'-[2-[(2Z)-1-(3,4-Dimethylphenyl)-1,5-dihydro-3-methyl-5-oxo-4H-pyrazol-4-ylidene]hydrazinyl]-2'-hydroxy-[1,1'-biphenyl]-3-carboxylic acid-13C4, SB 497115-13C4
LEDER [13CH3][13C](/[13C]([13C]1=O)=N/NC2=CC=CC(C3=CC(C(O)=O)=CC=C3)=C2O)=NN1C4=CC=C(C)C(C)=C4
Specifikationer
Kemiskt namn eller material Eltrombopag-13C4
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C25H22N4O4/c1-14-10-11-19(12-15(14)2)29-24(31)22(16(3)28-29)27-26-21-9-5-8-20(23(21)30)17-6-4-7-18(13-17)25(32)33/h4-13,28,30H,1-3H3,(H,32,33)/i3+1,16+1,22+1,24+1
Procent renhet >95%