Läs mer
Eltrombopag Methyl Ester, TRC
Produktkod. 30010311
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010311 50 mg
30006693 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010311 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE50800550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246929-01-0
Molekylformel C26H24N4O4
Molekylvikt (g/mol) 456.49
Synonym 3'-[(2Z)-2-[1-(3,4-Dimethylphenyl)-1,5-dihydro-3-methyl-5-oxo-4H-pyrazol-4-ylidene]hydrazinyl]-2'-hydroxy-[1,1'-Biphenyl]-3-carboxylic Acid Methyl Ester
LEDER O=C(N(N=C/1C)C2=CC=C(C)C(C)=C2)C1=N/NC3=CC=CC(C4=CC(C(OC)=O)=CC=C4)=C3O
Specifikationer
Kemiskt namn eller material Eltrombopag Methyl Ester
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C26H24N4O4/c1-15-11-12-20(13-16(15)2)30-25(32)23(17(3)29-30)28-27-22-10-6-9-21(24(22)31)18-7-5-8-19(14-18)26(33)34-4/h5-14,29,31H,1-4H3
Procent renhet >95%